4-(Dihydroxyboranyl)-2,3-difluorobenzoic acid 95+%
Catalog # APOSPC412389-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
Synonyms: 4-Borono-2,3-difluorobenzoic acid, 4-Carboxy-2,3-difluorophenylboronic acid
:
:201.92 g/mol
Storage Temperature:Ambient
CAS Number:1029716-92-4
Formula:C₇H₅BF₂O₄
MDL Number:MFCD06203343