[3-Cyclopropyl-4-(difluoromethyl)-6-(1,5-dimethyl-1H-pyrazol-4-yl)-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid
Catalog # APOSPC410487-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:361.346 g/mol
Storage Temperature:Ambient
CAS Number:1006478-10-9
Formula:C₁₇H₁₇F₂N₅O₂
MDL Number:MFCD08697750