Order Entry
France
ContactUsLinkComponent
[6-(1,5-Dimethyl-1H-pyrazol-4-yl)-3-methyl-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid
[6-(1,5-Dimethyl-1H-pyrazol-4-yl)-3-methyl-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid
Catalog # APOSPC410484-1G
Supplier:  Apollo Scientific
CAS Number:  
[6-(1,5-Dimethyl-1H-pyrazol-4-yl)-3-methyl-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid
Catalog # APOSPC410484-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :353.299 g/mol
Storage Temperature:Ambient
CAS Number:1006477-44-6
Formula:C₁₅H₁₄F₃N₅O₂
MDL Number:MFCD08697655