Order Entry
France
ContactUsLinkComponent
 
2-[4-Nitro-3-(trifluoromethyl)-1H-pyrazol-1-yl]butanoic acid
Catalog # APOSPC410460-1G
Supplier:  Apollo Scientific
CAS Number:  
2-[4-Nitro-3-(trifluoromethyl)-1H-pyrazol-1-yl]butanoic acid
Catalog # APOSPC410460-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :267.162 g/mol
Storage Temperature:Ambient
CAS Number:1006459-45-5
Formula:C₈H₈F₃N₃O₄
MDL Number:MFCD04970066