Order Entry
France
ContactUsLinkComponent
[3-Cyclopropyl-6-(1,5-dimethyl-1H-pyrazol-4-yl)-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid
[3-Cyclopropyl-6-(1,5-dimethyl-1H-pyrazol-4-yl)-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid
Catalog # APOSPC410445-1G
Supplier:  Apollo Scientific
CAS Number:  
[3-Cyclopropyl-6-(1,5-dimethyl-1H-pyrazol-4-yl)-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid
Catalog # APOSPC410445-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :379.336 g/mol
Storage Temperature:Ambient
CAS Number:1006444-33-2
Formula:C₁₇H₁₆F₃N₅O₂
MDL Number:MFCD08697673