Order Entry
France
ContactUsLinkComponent
3-Amino-6-(difluoromethyl)-4-(1,5-dimethyl-1H-pyrazol-4-yl)thieno[2,3-b]pyridine-2-carboxamide
3-Amino-6-(difluoromethyl)-4-(1,5-dimethyl-1H-pyrazol-4-yl)thieno[2,3-b]pyridine-2-carboxamide
Catalog # APOSPC410364-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Amino-6-(difluoromethyl)-4-(1,5-dimethyl-1H-pyrazol-4-yl)thieno[2,3-b]pyridine-2-carboxamide
Catalog # APOSPC410364-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :337.348 g/mol
Storage Temperature:Ambient
CAS Number:1005699-94-4
Formula:C₁₄H₁₃F₂N₅OS
MDL Number:MFCD03422785