3-Amino-4-(1-ethyl-3-methyl-1H-pyrazol-4-yl)-6-(trifluoromethyl)thieno[2,3-b]pyridine-2-carboxamide
Catalog # APOSPC410363-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:369.365 g/mol
Storage Temperature:Ambient
CAS Number:1005696-22-9
Formula:C₁₅H₁₄F₃N₅OS
MDL Number:MFCD03422769