3-Amino-6-(difluoromethyl)-4-(1,3-dimethyl-1H-pyrazol-4-yl)thieno[2,3-b]pyridine-2-carboxamide
Catalog # APOSPC410361-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:337.348 g/mol
Storage Temperature:Ambient
CAS Number:1005694-50-7
Formula:C₁₄H₁₃F₂N₅OS
MDL Number:MFCD03422784