Order Entry
France
ContactUsLinkComponent
3-Amino-6-(difluoromethyl)-4-(1-ethyl-3-methyl-1H-pyrazol-4-yl)thieno[2,3-b]pyridine-2-carboxamide
3-Amino-6-(difluoromethyl)-4-(1-ethyl-3-methyl-1H-pyrazol-4-yl)thieno[2,3-b]pyridine-2-carboxamide
Catalog # APOSPC410360-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Amino-6-(difluoromethyl)-4-(1-ethyl-3-methyl-1H-pyrazol-4-yl)thieno[2,3-b]pyridine-2-carboxamide
Catalog # APOSPC410360-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :351.374 g/mol
Storage Temperature:Ambient
CAS Number:1005679-22-0
Formula:C₁₅H₁₅F₂N₅OS
MDL Number:MFCD03422786