Order Entry
France
ContactUsLinkComponent
3-Amino-4-(1,5-dimethyl-1H-pyrazol-4-yl)-6-(trifluoromethyl)thieno[2,3-b]pyridine-2-carboxamide
3-Amino-4-(1,5-dimethyl-1H-pyrazol-4-yl)-6-(trifluoromethyl)thieno[2,3-b]pyridine-2-carboxamide
Catalog # APOSPC410352-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Amino-4-(1,5-dimethyl-1H-pyrazol-4-yl)-6-(trifluoromethyl)thieno[2,3-b]pyridine-2-carboxamide
Catalog # APOSPC410352-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :355.338 g/mol
Storage Temperature:Ambient
CAS Number:1005632-64-3
Formula:C₁₄H₁₂F₃N₅OS
MDL Number:MFCD03422767