1-(2,3-Difluorophenyl)-N-[(1,3-dimethyl-1H-pyrazol-4-yl)methyl]methanamine
Catalog # APOSPC410048-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:251.275 g/mol
Storage Temperature:Ambient
Formula:C₁₃H₁₅F₂N₃
MDL Number:MFCD08696807