2-(3-Fluoro-4-methoxy-benzyl)-piperidine hydrochloride
Catalog # APOSPC404532-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
:
:259.747 g/mol
Storage Temperature:Ambient
CAS Number:1170364-71-2
Formula:C₁₃H₁₈FNO·ClH
MDL Number:MFCD08669729