tert-Butyl (3-amino-2,6-difluorophenyl)carbamate
Catalog # APOSPC402000-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:244.238 g/mol
Storage Temperature:Ambient
CAS Number:535170-20-8
Formula:C₁₁H₁₄F₂N₂O₂
MDL Number:MFCD13182479