Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(3-Fluorophenyl)propylamine
1-(3-Fluorophenyl)propylamine
1-(3-Fluorophenyl)propylamine
APOSPC3484-250MG
:
Apollo Scientific
:
1-(3-Fluorophenyl)propylamine
APOSPC3484-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
153.197 g/mol
:
85 °C
:
GHS07
:
Ambient
:
473732-57-9
:
C₉H₁₂FN
:
MFCD07784284