6-Phenyl-4-(trifluoromethyl)thieno[2,3-b]pyridin-3-amine
Catalog # APOSPC300828-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:294.295 g/mol
Storage Temperature:Ambient
CAS Number:104960-57-8
Formula:C₁₄H₉F₃N₂S
MDL Number:MFCD28148251