N-(3,4-Difluorophenyl)-2-(1-methyl-1H-pyrrol-2-yl)-2-oxoacetamide, tech
Catalog # APOSPC28014-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:264.228 g/mol
Storage Temperature:Ambient
CAS Number:477872-04-1
Formula:C₁₃H₁₀F₂N₂O₂
MDL Number:MFCD02102458