Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
7-Fluoro-4-oxo-4H-chromene-2-carboxylic acid
7-Fluoro-4-oxo-4H-chromene-2-carboxylic acid 98%
7-Fluoro-4-oxo-4H-chromene-2-carboxylic acid 98%
APOSPC200548-250MG
:
Apollo Scientific
:
7-Fluoro-4-oxo-4H-chromene-2-carboxylic acid 98%
APOSPC200548-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
208.143 g/mol
:
Ambient
:
128942-39-2
:
C₁₀H₅FO₄
:
MFCD11877866