
N-[(2,3-Difluorophenyl)carbamothioyl]benzamide
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:887267-30-3
:
Formula:C₁₄H₁₀F₂N₂OS
MDL Number:MFCD08458007
:292.304 g/mol
Melting Pt:140…143 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together