Order Entry
France
ContactUsLinkComponent
3-(Nonafluoro-tert-butyl)propan-1-ol
3-(Nonafluoro-tert-butyl)propan-1-ol
Catalog # APOSPC10078-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(Nonafluoro-tert-butyl)propan-1-ol
Catalog # APOSPC10078-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Boiling Pt:134…135 °C
Melting Pt:36…37 °C
 :264.089 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:14115-49-2
Formula:C₆H₅F₉O
MDL Number:MFCD00459229