
4,4,4-Trifluorobutane-1-sulphonyl chloride
Catalog # APOSPC01483-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:210.602 g/mol
Storage Temperature:Ambient
UN:3265
ADR:8,III
:GHS05
CAS Number:212190-25-5
Formula:C₄H₆ClF₃O₂S
MDL Number:MFCD06657959