(3S)-1,2,3,4-Tetrahydroisoquinoline-7-hydroxy-3-carboxylic acid dihydrate 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:210709-23-2
:
Formula:C₁₀H₁₁NO₃
MDL Number:MFCD00819133
:193.199 g/mol
Storage Temperature:Ambient
Frequently Bought Together