Order Entry
France
ContactUsLinkComponent
1,3-Dihydro-2-benzofuran-5-carbaldehyde 95%
Irritant
1,3-Dihydro-2-benzofuran-5-carbaldehyde 95%
Catalog # APOSOR943816-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1,3-Dihydro-2-benzofuran-5-carbaldehyde 95%
Catalog # APOSOR943816-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :148.159 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:89424-83-9
Formula:C₉H₈O₂
MDL Number:MFCD09842492