Order Entry
France
ContactUsLinkComponent
4-Bromo-6-methylpyridin-2-ol 95%
4-Bromo-6-methylpyridin-2-ol 95%
Catalog # APOSOR928203-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Bromo-6-methylpyridin-2-ol 95%
Catalog # APOSOR928203-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :188.022 g/mol
Storage Temperature:Ambient
CAS Number:865156-59-8
Formula:C₆H₆BrNO
MDL Number:MFCD16610030