Order Entry
France
ContactUsLinkComponent
 
Trans-1,2-cyclopropane-dicarboxylic acid mono ethyl ester 95%
  APOSOR920382-1G
 :  
Trans-1,2-cyclopropane-dicarboxylic acid mono ethyl ester 95%
  APOSOR920382-1G
 :  

 

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

 

 

 :
 :158.152 g/mol
 :Ambient
 :31420-66-3
 :C₇H₁₀O₄
 :MFCD26383405