Order Entry
France
ContactUsLinkComponent
2-Isopropoxy-6-methylpyridine 96%
2-Isopropoxy-6-methylpyridine 96%
Catalog # APOSOR911168-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Isopropoxy-6-methylpyridine 96%
Catalog # APOSOR911168-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :151.206 g/mol
Storage Temperature:Ambient
CAS Number:1395039-68-5
Formula:C₉H₁₃NO
MDL Number:MFCD23114609