Order Entry
France
ContactUsLinkComponent
Thieno[2,3-b]pyridine-2-carboxaldehyde 97+%
Thieno[2,3-b]pyridine-2-carboxaldehyde 97+%
Catalog # APOSOR6738-100MG
Supplier:  Apollo Scientific
CAS Number:  
Thieno[2,3-b]pyridine-2-carboxaldehyde 97+%
Catalog # APOSOR6738-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :163.196 g/mol
Melting Pt:141.5…143 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:53174-98-4
Formula:C₈H₅NOS
MDL Number:MFCD07772791