Order Entry
France
ContactUsLinkComponent
2-(Aminomethyl)-3-bromo-5-methylpyridine hydrochloride
2-(Aminomethyl)-3-bromo-5-methylpyridine hydrochloride
Catalog # APOSOR60142-100MG
Supplier:  Apollo Scientific
CAS Number:  
2-(Aminomethyl)-3-bromo-5-methylpyridine hydrochloride
Catalog # APOSOR60142-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440
  • Environmentally Preferable:

Specifications

About this item

Synonyms: (3-Bromo-5-methylpyridin-2-yl)methylamine hydrochloride
 :
 :237.525 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:1257535-62-8
Formula:C₇H₉BrN₂·ClH
MDL Number:MFCD17214207