Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(7-Bromonaphthalen-1-yl)cyclopropan-1-amine
1-(7-Bromonaphthalen-1-yl)cyclopropan-1-amine
1-(7-Bromonaphthalen-1-yl)cyclopropan-1-amine
APOSOR480166-1G
:
Apollo Scientific
:
1-(7-Bromonaphthalen-1-yl)cyclopropan-1-amine
APOSOR480166-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
262.145 g/mol
:
Ambient
:
1785577-54-9
:
C₁₃H₁₂BrN
:
MFCD30496538