1-(2-Bromonaphthalen-1-yl)cyclopropane-1-carboxylic acid
Catalog # APOSOR480139-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:291.14 g/mol
Storage Temperature:Ambient
Formula:C₁₄H₁₁BrO₂
MDL Number:MFCD30496578