Order Entry
France
ContactUsLinkComponent
 
5-(2-Methoxyphenyl)nicotinic acid
Catalog # APOSOR470407-5G
Supplier:  Apollo Scientific
CAS Number:  
5-(2-Methoxyphenyl)nicotinic acid
Catalog # APOSOR470407-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :229.231 g/mol
Storage Temperature:Ambient
CAS Number:893732-61-1
Formula:C₁₃H₁₁NO₃
MDL Number:MFCD06410272