Order Entry
France
ContactUsLinkComponent
5-(4-Bromophenyl)nicotinic acid
5-(4-Bromophenyl)nicotinic acid
Catalog # APOSOR470330-5G
Supplier:  Apollo Scientific
CAS Number:  
5-(4-Bromophenyl)nicotinic acid
Catalog # APOSOR470330-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :278.101 g/mol
Storage Temperature:Ambient
CAS Number:887973-36-6
Formula:C₁₂H₈BrNO₂
MDL Number:MFCD06410263