1-(2-Methoxyethyl)-2-oxopyrrolidine-4-carboxamidine hydrochloride
Catalog # APOSOR470182-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:221.685 g/mol
Storage Temperature:Ambient
CAS Number:1272756-21-4
Formula:C₈H₁₅N₃O₂·ClH
MDL Number:MFCD18651698