1-Isopropyl-2-oxopyrrolidine-4-carboxamidine hydrochloride
Catalog # APOSOR470181-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:205.685 g/mol
Storage Temperature:Ambient
CAS Number:1272756-01-0
Formula:C₈H₁₅N₃O·ClH
MDL Number:MFCD18651697