
3-Bromo-2-nitrobenzenesulphonyl chloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1261671-62-8
:
Formula:C₆H₃BrClNO₄S
MDL Number:MFCD18392272
:300.514 g/mol
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
Frequently Bought Together