4-Bromo-5-methyl-2-nitrophenol
Catalog # APOSOR400198-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:232.031 g/mol
Storage Temperature:Ambient
CAS Number:182500-28-3
Formula:C₇H₆BrNO₃
MDL Number:MFCD12923347