Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Benzyloxy-2-methylpyridine
5-Benzyloxy-2-methylpyridine 95+%
5-Benzyloxy-2-methylpyridine 95+%
5-Benzyloxy-2-methylpyridine 95+%
APOSOR346578-1G
:
Apollo Scientific
:
5-Benzyloxy-2-methylpyridine 95+%
APOSOR346578-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
199.248 g/mol
:
Ambient
:
63793-98-6
:
C₁₃H₁₃NO
:
MFCD15523327