Order Entry
France
ContactUsLinkComponent
4-Iodo-2-methylpyridine 98%
4-Iodo-2-methylpyridine 98%
Catalog # APOSOR345298-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Iodo-2-methylpyridine 98%
Catalog # APOSOR345298-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :219.023 g/mol
Storage Temperature:Ambient
CAS Number:22282-65-1
Formula:C₆H₆IN
MDL Number:MFCD09702484