Order Entry
France
ContactUsLinkComponent
(R)-3,3'-Bis[3,5-dimethylphenyl]-1,1'-binapthyl-2,2'-diyl hydrogenphosphate
(R)-3,3'-Bis[3,5-dimethylphenyl]-1,1'-binapthyl-2,2'-diyl hydrogenphosphate
Catalog # APOSOR340018-250MG
Supplier:  Apollo Scientific
CAS Number:  
(R)-3,3'-Bis[3,5-dimethylphenyl]-1,1'-binapthyl-2,2'-diyl hydrogenphosphate
Catalog # APOSOR340018-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :556.587 g/mol
Storage Temperature:Ambient
CAS Number:861909-53-7
Formula:C₃₆H₂₉O₄P
MDL Number:MFCD27952571