1-[5-(4-Methylpiperazin-1-yl)thiophen-2-yl]ethan-1-one, tech
Catalog # APOSOR32919-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:224.323 g/mol
Storage Temperature:Ambient
CAS Number:685107-45-3
Formula:C₁₁H₁₆N₂OS
MDL Number:MFCD03617419