(3Z)-3-[(4-Hydroxy-3-methoxy-5-methylphenyl)methylidene]-2,3-dihydro-1H-indol-2-one, tech
Catalog # APOSOR32142-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
:
:281.306 g/mol
Storage Temperature:Ambient
CAS Number:860648-89-1
Formula:C₁₇H₁₅NO₃
MDL Number:MFCD05668727