Propan-2-yl 5-bromothiophene-2-carboxylate
Catalog # APOSOR310417-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:249.125 g/mol
Storage Temperature:Ambient
CAS Number:1174013-07-0
Formula:C₈H₉BrO₂S
MDL Number:MFCD21101334