cis-1-Amino-1,2,3,4-tetrahydro-2-naphthalenecarboxylic acid
Catalog # APOSOR309433-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:191.226 g/mol
Storage Temperature:Ambient
CAS Number:888407-44-1
Formula:C₁₁H₁₃NO₂
MDL Number:MFCD09260408