Order Entry
France
ContactUsLinkComponent
3-tert-Butoxycarbonylamino-3-(3-nitro-phenyl)-propionic acid
3-tert-Butoxycarbonylamino-3-(3-nitro-phenyl)-propionic acid
Catalog # APOSOR307578-1G
Supplier:  Apollo Scientific
CAS Number:  
3-tert-Butoxycarbonylamino-3-(3-nitro-phenyl)-propionic acid
Catalog # APOSOR307578-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :310.303 g/mol
Storage Temperature:Ambient
CAS Number:284492-22-4
Formula:C₁₄H₁₈N₂O₆
MDL Number:MFCD02090669