
cis-4-Carbomethoxy-cyclohexane-1-carboxylic acid 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1011-85-4
:
Formula:C₉H₁₄O₄
MDL Number:MFCD01311246
:186.205 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together