
1-(3-Methyl-1,2,4-oxadiazol-5-yl)cyclopropan-1-amine hydrochloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1159737-11-7
:
Formula:C₆H₉N₃O·ClH
MDL Number:MFCD21602189
:175.616 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together