Pyrrolidine-1-carboxamidine hydrochloride
Catalog # APOSOR300903-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:149.622 g/mol
Storage Temperature:Ambient
CAS Number:49755-46-6
Formula:C₅H₁₁N₃·ClH
MDL Number:MFCD25963774