4-Benzylpiperidine-1-carboxamidine acetate
Catalog # APOSOR300898-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:277.362 g/mol
Storage Temperature:Ambient
CAS Number:1672675-23-8
Formula:C₁₃H₁₉N₃·CH₃CO₂H
MDL Number:MFCD28122846