


N,N-Dimethyl-1H-pyrrolo[2,3-c]pyridin-7-amine
Catalog # APOSOR300210-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:161.204 g/mol
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS06
:GHS05
:GHS07
CAS Number:2103352-79-8
Formula:C₉H₁₁N₃
MDL Number:MFCD31536734