2-(tert-Butyl)-6-(chloromethyl)pyrimidin-4-ol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:94171-08-1
:
Formula:C₉H₁₃ClN₂O
MDL Number:MFCD00084911
:200.665 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together