Benzo[b]thiophene-3-carboxamidine hydrochloride hydrate
Catalog # APOSOR23181-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:230.714 g/mol
Melting Pt:230 °C
:GHS07
Storage Temperature:Ambient
CAS Number:465515-36-0
Formula:C₉H₈N₂S·ClH·H₂O
MDL Number:MFCD03659730